Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT7 Monoclonal Antibody (OTI3B7), TrueMAB™, OriGene
SDP

Catalog No. 501679559
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9559 Supplier OriGene Supplier No. TA803065
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).

Specifications

Antigen KRT7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KRT7
Gene Accession No. P08729
Gene Alias CK7, K2C7, K7, SCL
Gene Symbols KRT7
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-114 and 340-469 of human KRT7 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.