Learn More
KRT7 Mouse anti-Human, Clone: OTI2D1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803332

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).Specifications
KRT7 | |
Monoclonal | |
Unconjugated | |
KRT7 | |
CK7, K2C7, K7, SCL | |
Mouse | |
Affinity Chromatography | |
RUO | |
3855 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2D1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08729 | |
KRT7 | |
Human recombit protein fragment corresponding to amino acids 1-114 and 340-469 of human KRT7 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.