Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT7 Mouse anti-Human, Unconjugated, Clone: KRT7/760 + OV-TL12/3, Abnova™

Catalog No. 89337767 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-767 Supplier Abnova Corporation Supplier No. MAB21276
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KRT7 Monoclonal antibody specifically detects KRT7 in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen KRT7
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone KRT7/760 + OV-TL12/3
Conjugate Unconjugated
Description keratin 7
Gene KRT7
Gene Alias CK7, K2C7, K7, MGC129731, MGC3625, SCL
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human KRT7
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.