Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT7, Rabbit anti-Human, Clone: R17-S, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15941
Specifications
KRT7 | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (20 mg/mL BSA, 0.05% Sodium Azide). | |
CK7/K2C7/K7/MGC129731/MGC3625/SCL | |
Rabbit | |
200 μL | |
Primary | |
Human | |
Antibody |
Immunohistochemistry (Paraffin) | |
R17-S | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:200) The optimal working dilution should be determined by the end user. | |
keratin 7 | |
KRT7 | |
A synthetic peptide corresponding to N-terminus of human KRT7. | |
RUO | |
3855 | |
Store at 4°C. Do not freeze. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction