Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT71 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583965 Supplier Invitrogen™ Supplier No. PA583965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFPSPVSISI ISSTSGGSGY GFRPSMVSGG YVANSSNCIS GVCSVRGGEG RSRGSANDYK DTLGKGSSLS APSKKTSR Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 82%.

K6IRS1 belongs to a family of type II keratins that are specifically expressed in the inner root sheath of hair follicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT71
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT71
Gene Accession No. Q3SY84
Gene Alias CK-71; cytokeratin-71; hK6irs; hK6irs1; HYPT13; K6IRS1; K71; KB34; Keratin 6 irs; keratin 71; keratin 71, type II; keratin, type II cytoskeletal 71; Keratin-71; KRT6IRS; KRT6IRS1; KRT71; Type II inner root sheath-specific keratin-K6irs1; type-II keratin Kb34
Gene Symbols KRT71
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT71. Recombinant protein control fragment (Product #RP-97765).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.