Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT71 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA528558

Catalog No. PIPA528558


Only null left
Add to Cart

Description

Description

Recommended positive controls: HepG2. Predicted reactivity: Mouse (91%), Rat (91%), Dog (91%), Cat (93%), Sheep (91%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

K6IRS1 belongs to a family of type II keratins that are specifically expressed in the inner root sheath of hair follicles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KRT71
Polyclonal
Unconjugated
KRT71
CK-71; cytokeratin-71; hK6irs; hK6irs1; HYPT13; K6IRS1; K71; KB34; Keratin 6 irs; keratin 71; keratin 71, type II; keratin, type II cytoskeletal 71; Keratin-71; KRT6IRS; KRT6IRS1; KRT71; Type II inner root sheath-specific keratin-K6irs1; type-II keratin Kb34
Rabbit
Antigen affinity chromatography
RUO
112802
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.91 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q3SY84
KRT71
Recombinant protein encompassing a sequence within the center region of human KRT71. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.