Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584694
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584694 Supplier Invitrogen™ Supplier No. PA584694
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSGGIGSSSA SFRARVKGSA SFGSKSLSCL GGSRSLALS Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 62%.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells. The type II keratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This gene encodes a type II keratin that is specifically expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q12-q13. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT72
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT72
Gene Accession No. Q14CN4
Gene Alias CK-72; cytokeratin-72; K6irs; K6IRS2; K72; KB35; keratin 6, inner root sheath, 2; keratin 72; keratin 72, type II; keratin protein K6irs; keratin, type II cytoskeletal 72; Keratin-72; KRT6; KRT6IRS2; KRT72; Type II inner root sheath-specific keratin-K6irs2; type-II keratin Kb35
Gene Symbols KRT72
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT72. Recombinant protein control fragment (Product #RP-107437).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.