Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT72 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530898
Description
Recommended positive controls: NT2D1. Predicted reactivity: Human (99%), Mouse (84%), Rat (83%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells. The type II keratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This gene encodes a type II keratin that is specifically expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q12-q13. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
KRT72 | |
Polyclonal | |
Unconjugated | |
KRT72 | |
CK-72; cytokeratin-72; K6irs; K6IRS2; K72; KB35; keratin 6, inner root sheath, 2; keratin 72; keratin 72, type II; keratin protein K6irs; keratin, type II cytoskeletal 72; Keratin-72; KRT6; KRT6IRS2; KRT72; Type II inner root sheath-specific keratin-K6irs2; type-II keratin Kb35 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140807 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q14CN4 | |
KRT72 | |
Recombinant protein fragment corresponding to a region within amino acids 217 and 434 of Human KRT72. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction