Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT73 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598581
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598581 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598581 Supplier Invitrogen™ Supplier No. PA598581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRT73 Polyclonal Antibody for Western Blot, ELISA

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes a protein that is expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT73
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KRT73
Gene Accession No. Q86Y46
Gene Alias BC067067; CK-73; cytokeratin-73; IRT6IRS3; K6IRS3; K73; Kb36; keratin 6 irs3; keratin 73; keratin 73, type II; keratin, type II cytoskeletal 73; Keratin-73; KRT6IRS3; Krt73; Type II inner root sheath-specific keratin-K6irs3; type II keratin Kb36; type-II keratin Kb36
Gene Symbols KRT73
Host Species Rabbit
Immunogen Recombinant Human Keratin, type II cytoskeletal 73 protein (1-300AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319101
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.