Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT74 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144792 Supplier Invitrogen™ Supplier No. PA5144792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISVISSSSYS YHHPSSAGVD LGASAVAGSS GSTQSGQTKT TEARGGDLKD TQGKSTPASI PARKATR Antigen sequence identity: Mouse 36%; Rat 36%.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This protein belongs to a family of keratins that are specifically expressed in the inner root sheath of hair follicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT74
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT74
Gene Accession No. Q7RTS7
Gene Alias ADWH; CK-74; cytokeratin-74; HTSS2; HYPT3; K5C; K6IRS4; K74; KB37; keratin 5c; keratin 6 irs4; keratin 74; keratin 74, type II; keratin, type II cytoskeletal 74; Keratin-5c; keratin-74; KRT5C; KRT6IRS4; KRT74; Type II inner root sheath-specific keratin-K6irs4; type-II keratin Kb37
Gene Symbols KRT74
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 463-529. Recombinant protein control fragment (Product #RP-97821).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121391
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.