Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT76, anti-Human, Clone: KRTH/1576R, Abnova™
Monoclonal Antibody
Supplier: Abnova Corporation RAB00379
Specifications
KRT76 | |
Monoclonal | |
Unconjugated | |
In 10 mM PBS (0.05% BSA, 0.05% sodium azide). | |
HUMCYT2A/KRT2B/KRT2P | |
Original antibody is raised against recombinant protein corresponding to full length human KRT76. | |
RUO | |
51350 | |
Store at 4°C. | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
KRTH/1576R | |
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
keratin 76 | |
KRT76 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction