Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT76 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546516
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546516 Supplier Invitrogen™ Supplier No. PA546516
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GSISVSHSGM GSSSGSIQTS GGSGYKSGGG GSTSIRFSQT TSSSQHSSTK Sequence homology: Dog: 92%; Horse: 100%; Human: 100%; Mouse: 83%; Rat: 83%.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT76
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KRT76
Gene Accession No. Q01546
Gene Alias 2310001L23Rik; CK-2P; cytokeratin 2; cytokeratin-2P; HUMCYT2A; K2P; K76; Kb9; keratin 2p; keratin 76; keratin 76, type II; keratin, type II cytoskeletal 2 oral; Keratin-76; KRT2B; KRT2P; KRT76; masticatory epithelia keratin 2p; type II keratin Kb9; type-II keratin Kb9
Gene Symbols KRT76
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KRT76 (aa 589-638).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51350
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.