Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT76 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554046
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554046 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554046 Supplier Invitrogen™ Supplier No. PA554046
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGSGYGGVSS GSTGGRGSSG SYQSSSSGSR LGGAGSISVS HSGMGSSSGS IQTSGGSGYK SGGGGSTSIR FSQTTSSSQH SSTK Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 57%.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT76
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT76
Gene Accession No. Q01546
Gene Alias 2310001L23Rik; CK-2P; cytokeratin 2; cytokeratin-2P; HUMCYT2A; K2P; K76; Kb9; keratin 2p; keratin 76; keratin 76, type II; keratin, type II cytoskeletal 2 oral; Keratin-76; KRT2B; KRT2P; KRT76; masticatory epithelia keratin 2p; type II keratin Kb9; type-II keratin Kb9
Gene Symbols KRT76
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT76. Recombinant protein control fragment (Product #RP-92536).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51350
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.