Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT76 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: KRTH/2147R, Abnova™

Catalog No. 89339730 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-730 Supplier Abnova Corporation Supplier No. RAB00691
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

KRT76 Monoclonal antibody specifically detects KRT76 in Human, Mouse, Rat samples. It is validated for WB, IHC-P, IF, Flow Cyt.

Specifications

Antigen KRT76
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone KRTH/2147R
Conjugate Unconjugated
Description keratin 76
Gene KRT76
Gene Alias HUMCYT2A, KRT2B, KRT2P
Host Species Rabbit
Immunogen Solubilized keratin extract from human stratum corneum
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 51350
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.