Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT77 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100848 Supplier Invitrogen™ Supplier No. PA5100848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total K2C1B.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. KRT77 encodes an epithelial keratin that is expressed in the skin and eccrine sweat glands. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT77
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KRT77
Gene Accession No. Q6IG01, Q7Z794
Gene Alias 4732484G22Rik; CK-1B; CK-2P; cytokeratin 2; cytokeratin-1B; Cytokeratin-2P; embryonic type II keratin 1; embryonic type II keratin-1; HUMCYT2A; K1B; K1emb; K2P; K76; K77; Kb39; keratin 1b; keratin 2p; keratin 76; keratin 76, type II; keratin 77; keratin 77, type II; keratin complex 2, basic, gene 39; keratin, type II cytoskeletal 1b; Keratin, type II cytoskeletal 2 oral; keratin-76; Keratin-77; Krt1b; Krt239; KRT2B; KRT2P; KRT76; Krt77; mK1b; type II keratin Kb39; type-II intermediate filament protein; type-II keratin Kb39; Type-II keratin Kb9
Gene Symbols KRT77
Host Species Rabbit
Immunogen A synthesized peptide derived from human KRT77(Accession Q7Z794), corresponding to amino acid residues L375-W425.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374454, 406226
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.