Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT77 Rabbit anti-Human, Unconjugated, Clone: KRTH/1576R + KRTL/15, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation RAB00689
Description
KRT77 Monoclonal antibody specifically detects KRT77 in Human samples. It is validated for IHC-P.Specifications
KRT77 | |
Monoclonal | |
Unconjugated | |
KRT77 | |
Rabbit | |
100 μg | |
374454, 51350 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunohistochemistry (Paraffin) | |
KRTH/1576R + KRTL/15 | |
keratin 77 | |
K1B, KRT1B, MGC148087 | |
Recombinant protein corresponding to full-length human KRT77 and KRT76 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction