Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT78 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558902 Supplier Invitrogen™ Supplier No. PA558902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGGGLGSTCG LGSGKGSPGS CCTSIVTGGS NIILGSGKDP VLDSCSVSGS S Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 52%.

This gene is a member of the type II keratin gene family and encodes a protein with an intermediate filament domain. Keratins are the major structural proteins in epithelial cells, forming a cytoplasmic network of 10 to 12 nm wide intermediate filaments and creating a scaffold that gives cells the ability to withstand mechanical and non-mechanical stresses. The genes of the type II keratin family are located as a gene cluster at 12p13.13. Four pseudogenes of this gene family have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT78
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT78
Gene Accession No. Q8N1N4
Gene Alias CK-78; Cytokeratin-78; K5B; K78; Kb40; keratin 5b; keratin 78; keratin 78, type II; keratin, type II cytoskeletal 78; Keratin-5b; Keratin-78; KRT78; Type-II keratin Kb40
Gene Symbols KRT78
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT78. Recombinant protein control fragment (Product #RP-97817).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196374
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.