Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT79 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563706
Description
Immunogen sequence: MRSSVSRQTY STKGGFSSNS ASGGSGSQAR TSFSSVTVSR SSGSGGGAHC GPGTGGFGS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 47%.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes an epithelial keratin that is expressed in skeletal muscle, skin and scalp. The type II keratins are clustered in a region of chromosome 12q13.
Specifications
| KRT79 | |
| Polyclonal | |
| Unconjugated | |
| KRT79 | |
| BC031593; CK-79; cytokeratin-79; K6L; K79; KB38; keratin 6L; keratin 79; keratin 79, type II; keratin, type II cytoskeletal 79; Keratin-6L; Keratin-6-like; keratin-79; KRT6L; KRT79; Type-II keratin Kb38 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 338785 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XKE5 | |
| KRT79 | |
| Recombinant protein corresponding to Human KRT79. Recombinant protein control fragment (Product #RP-97874). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction