Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT79 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA581209

Catalog No. PIPA581209


Only null left
Add to Cart

Description

Description

This antibody has specificity for Human KRT6L/KRT79.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes an epithelial keratin that is expressed in skeletal muscle, skin and scalp. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KRT79
Polyclonal
Unconjugated
KRT79
BC031593; CK-79; cytokeratin-79; K6L; K79; KB38; keratin 6L; keratin 79; keratin 79, type II; keratin, type II cytoskeletal 79; Keratin-6L; Keratin-6-like; keratin-79; KRT6L; KRT79; Type-II keratin Kb38
Rabbit
Antigen affinity chromatography, Protein A
RUO
338785
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.03% ProClin 300; pH 7
Q5XKE5
KRT79
A synthetic peptide corresponding to the center region of the Human KRT6L/KRT79.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.