Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT79 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546517 Supplier Invitrogen™ Supplier No. PA546517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EVKAQYELIA QRSRAEAEAW YQTKYEELQV TAGKHGDNLR DTKNEIAELT Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 79%; Rat: 100%.

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes an epithelial keratin that is expressed in skeletal muscle, skin and scalp. The type II keratins are clustered in a region of chromosome 12q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT79
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KRT79
Gene Accession No. Q5XKE5
Gene Alias BC031593; CK-79; cytokeratin-79; K6L; K79; KB38; keratin 6L; keratin 79; keratin 79, type II; keratin, type II cytoskeletal 79; Keratin-6L; Keratin-6-like; keratin-79; KRT6L; KRT79; Type-II keratin Kb38
Gene Symbols KRT79
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human KRT79 (aa 317-366).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338785
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.