Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT8 Mouse anti-Human, Unconjugated, Clone: rB22.1, Abnova™

Catalog No. 89337929 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-929 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-929 Supplier Abnova Corporation Supplier No. RAB00504
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KRT8 Monoclonal antibody specifically detects KRT8 in Human samples. It is validated for IHC-P.

Specifications

Antigen KRT8
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rB22.1
Conjugate Unconjugated
Description keratin 8
Gene KRT8
Gene Alias CARD2, CK8, CYK8, K2C8, K8, KO
Host Species Mouse
Immunogen Original antibody is raised against recombinant protein corresponding to full length human KRT8
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3856
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.