Learn More
Invitrogen™ KRT80 Polyclonal Antibody

Description
Immunogen sequence: GEEGRMDSPSA TVVSAVQSRC KTAASRSGLS KAPSRKKKGS KGPVIKITEM SEKYFSQESE VSE Highest antigen sequence identity to the following orthologs - mouse 75%, rat 81%.
Specifications
Specifications
| Antigen | KRT80 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KRT80 |
| Gene Accession No. | Q6KB66 |
| Gene Alias | 1200016G03Rik; 2310041I20Rik; CK-80; cytokeratin-80; K80; Kb2; KB20; keratin 80; keratin 80, type II; keratin b20; keratin, type II cytoskeletal 80; Keratin-80; KRT80; type II keratin; type II keratin Kb20; type II keratin-20; type-II keratin Kb20 |
| Gene Symbols | KRT80 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.