Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT80 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598537
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598537 Supplier Invitrogen™ Supplier No. PA598537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRT80 Polyclonal Antibody for ELISA, IHC (P), Western Blot

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene's expression profile shows that it encodes a type II epithelial keratin, although structurally the encoded protein is more like a type II hair keratin. This protein is involved in cell differentiation, localizing near desmosomal plaques in earlier stages of differentiation but then dispersing throughout the cytoplasm in terminally differentiating cells. The type II keratins are clustered in a region of chromosome 12q13. Two transcript variants encoding two different fully functional isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT80
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KRT80
Gene Accession No. Q0VBK2, Q6KB66
Gene Alias 1200016G03Rik; 2310041I20Rik; CK-80; cytokeratin-80; K80; Kb2; KB20; keratin 80; keratin 80, type II; keratin b20; keratin, type II cytoskeletal 80; Keratin-80; KRT80; type II keratin; type II keratin Kb20; type II keratin-20; type-II keratin Kb20
Gene Symbols KRT80
Host Species Rabbit
Immunogen Recombinant Human Keratin, type II cytoskeletal 80 protein (1-300AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144501, 74127
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.