Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592102
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592102 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592102 Supplier Invitrogen™ Supplier No. PA592102
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAQSEQQGEA ALSDARCKLA ELEGALQKAK QDMACLIREY QEVMNSKLGL DIEIATYRRL LEGEEQRLCE GIGAVNVCVS SSRGGVVCGD LCVSGSRPVT G Positive Samples: Raji, A-549.

The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this hair keratin, as well as KRTHB3 and KRTHB6, is found primarily in the hair cortex. Mutations in this gene and KRTHB6 have been observed in patients with a rare dominant hair disease, monilethrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT81
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KRT81
Gene Accession No. Q14533, Q9ERE2
Gene Alias AA589625; ghHb1; ghHkb1; Hair keratin K2.9; hard keratin, type II, 1; HB1; Hb-1; hHAKB2-1; K81; Kb21; keratin 81; keratin 81, type II; keratin complex 2, basic, gene 19; keratin, hair, basic, 1; Keratin, type II cuticular Hb1; keratin-81; Krt2-19; KRT81; KRTHB1; Metastatic lymph node 137 gene protein; MLN 137; MLN137; type II hair keratin Hb1; type II keratin Kb21; type-II keratin Kb21
Gene Symbols KRT81
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human KRT81 (NP_0022722).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3887, 64818
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.