Learn More
Invitrogen™ KRT84 Monoclonal Antibody (OTI2D1)

Description
KRT84 Monoclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | KRT84 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI2D1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | KRT84 |
| Gene Accession No. | Q9NSB2 |
| Gene Alias | 65 kDa type II keratin; AV235125; hard keratin, type II, 4; HB4; HRb-1; K84; Kb24; keratin 84; keratin 84, type II; keratin complex 2, basic, gene 16; keratin complex 2, gene 16; keratin, hair, basic, 4; keratin, type II cuticular Hb4; Keratin-84; Krt2-16; Krt2-3; Krt84; KRTHB4; type II hair keratin 4; type II hair keratin Hb4; type II keratin Kb24; type-II keratin Kb24 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.