Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT85 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570733
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570733 Supplier Thermo Scientific Supplier No. PA570733
Add to Cart
Add to Cart

KRT85 Polyclonal Antibody

KRT85 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT85
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KRT85
Gene Accession No. P78386
Gene Alias AA589597; BB007925; ECTD4; hair keratin; Hair keratin K2.12; hard keratin, type II, 5; HB5; Hb-5; hHb5; K85; Kb25; keratin 85; keratin 85, type II; keratin 89; keratin complex 2, basic, gene 18; keratin, hair, basic, 5; keratin, type II cuticular Hb5; Keratin-85; Krt2-18; Krt85; KRT89; Krthb5; Type II hair keratin Hb5; type II keratin Kb25; type-II keratin Kb25
Gene Symbols KRT85
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human KRT85.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3891
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.