Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT87P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572087
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572087 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572087 Supplier Invitrogen™ Supplier No. PA572087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRT87P Polyclonal Antibody for Western Blot

KRT87P (Keratin 87 Pseudogene) is a Pseudogene. Gene Ontology (GO) annotations related to this gene includestructural molecule activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT87P
Applications Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene KRT87P
Gene Accession No. A6NCN2
Gene Alias HBD; keratin 87 pseudogene; KRT121P; KRT87P; KRTHBP4
Gene Symbols KRT87P
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 96-122 amino acids from the N-terminal region of human KT81L.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.