Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587647 Supplier Invitrogen™ Supplier No. PA587647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TANEKSTMQE LNSRLASYLD KVQALEEANN DLENKIQDWY DKKGPAAIQK NYSPYYNTID DLKDQIVDLT VGNNKTLLDI DNTRMTLDDF RIKFEMEQNL RQGVDADING LRQVLDNLTM EKSDLEMQYE TLQEELMALK KNHKEEMSQL TGQNSGDVNV EINVAPGKDL TKTLNDMRQE YEQLIAKNRK DIENQYETQI TQIEHEVSSS GQEVQSSAKE VTQLRHGVQE LEIELQSQLS KKAALEKSLE DTKNRYCGQL QMIQEQISNL EAQITDVRQE IECQNQEYSL LLSIKMRLEK E; Positive Samples: HeLa, LO2, U-251MG, Mouse liver, Mouse testis, Rat testis.

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT9
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KRT9
Gene Accession No. P35527, Q6RHW0
Gene Alias CK-9; cytokeratin 9; cytokeratin-9; EPPK; K9; keratin 9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 9; keratin, type I cytoskeletal 9; Keratin-9; Krt1-9; Krt9; spermatid perinuclear ring manchette protein K9; type I cytoskeletal 9
Gene Symbols KRT9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 150-450 of human Cytokeratin 9 (KRT9) (KRT9) (NP_0002172).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107656, 3857
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.