Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT9 Rabbit anti-Human, Clone: SD201-09, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267810
Description
KRT9 Monoclonal specifically detects KRT9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KRT9 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
cytokeratin-9, EPPK, keratin 9, keratin, type I cytoskeletal 9 | |
Rabbit | |
Protein A purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
SD201-09 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
KRT9 | |
Recombinant protein wiehin Human KRT9 aa 234-422/ 623. (SwissProt: P35527 Human; SwissProt: Q6RHW0 Mouse; SwissProt: Q8CIS9 Rat) | |
100 μL | |
3857 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction