Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT9 Rabbit anti-Human, Mouse, Rat, Clone: 9A5H3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | KRT9 |
---|---|
Clone | 9A5H3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
KRT9 Monoclonal antibody specifically detects KRT9 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
KRT9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3857 | |
IgG | |
Affinity purified |
9A5H3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
cytokeratin-9, EPPK, keratin 9, keratin, type I cytoskeletal 9 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 254-412 of human KRT9 (P35527). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title