Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRTAP1-1 Polyclonal Antibody

KRTAP1-1 Polyclonal Antibody
Supplier: Thermo Scientific PA525474
Description
KRTAP1-1 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
| KRTAP1-1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q07627 | |
| KRTAP1-1 | |
| KLH conjugated synthetic peptide between 88-117 amino acids from the central region of human KRTAP1-1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| KRTAP1-1 | |
| B2A; HB2A; high sulfur keratin-associated protein 1.1; hKAP1.7; KAP1.1; KAP1.1A; KAP1.1B; KAP1.6; KAP1.7; keratin associated protein 1.7; keratin associated protein 1-1; keratin associated protein 1A; Keratin-associated protein 1.1; keratin-associated protein 1.6; Keratin-associated protein 1.7; keratin-associated protein 1-1; KRTAP1.1; KRTAP1-1; KRTAP1A | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 81851 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction