Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP1-5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570493 Supplier Invitrogen™ Supplier No. PA570493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 92%; Goat: 92%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 86%; Rabbit: 92%; Rat: 92%; Sheep: 92%.

This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the high sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP1-5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KRTAP1-5
Gene Accession No. Q9BYS1
Gene Alias 2310043L02Rik; AI592983; high sulfur keratin-associated protein 1.5; high sulfur protein B2F; KAP1.5; keratin associated protein 1.5; keratin associated protein 1-5; Keratin-associated protein 1.5; keratin-associated protein 1-5; KRTAP1.5; KRTAP1-5
Gene Symbols KRTAP1-5
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human KRTAP1-5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83895
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.