Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRTAP11-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567559
Description
KRTAP11-1 Polyclonal Antibody for Western Blot
In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.
Specifications
KRTAP11-1 | |
Polyclonal | |
Unconjugated | |
KRTAP11-1 | |
AA162070; AA589467; AV018433; Hacl1; HACL-1; High sulfur keratin-associated protein 11.1; KAP11.1; keratin associated protein 11-1; keratin-associated protein 11.1; keratin-associated protein 11-1; KRTAP11.1; KRTAP11-1; PRED29 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16693, 337880 | |
-20°C | |
Liquid |
Western Blot | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IUC1 | |
KRTAP11-1 | |
Synthesized peptide derived from human KRTAP11-1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction