Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP11-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531957 Supplier Invitrogen™ Supplier No. PA531957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP11-1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene KRTAP11-1
Gene Accession No. Q8IUC1
Gene Alias AA162070; AA589467; AV018433; Hacl1; HACL-1; High sulfur keratin-associated protein 11.1; KAP11.1; keratin associated protein 11-1; keratin-associated protein 11.1; keratin-associated protein 11-1; KRTAP11.1; KRTAP11-1; PRED29
Gene Symbols KRTAP11-1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KRTAP11-1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 337880
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.