Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP16-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567136 Supplier Invitrogen™ Supplier No. PA567136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACVTSYSCRPV YFRPSCTESD SCKRDCKKST SSQLDCVDTT PCKVDVSEEA PCQPTEAKPI SPTTR Highest antigen sequence identity to the following orthologs - mouse 61%, rat 62%.

KRTAP16-1 gene ontology annotations related to this gene include keratinization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP16-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRTAP16-1
Gene Accession No. A8MUX0
Gene Alias KAP16.1; keratin associated protein 16-1; keratin-associated protein 16-1; KRTAP16-1; putative keratin-associated protein 10-like ENSP00000375147
Gene Symbols KRTAP16-1
Host Species Rabbit
Immunogen Recombinant Human KRTAP16-1. Recombinant protein control fragment (Product #RP-107781).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100505753
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.