Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRTAP2-4 Antibody (OTI3B4), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272304PECY7
Description
KRTAP2-4 Monoclonal specifically detects KRTAP2-4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
KRTAP2-4 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
KRTAP2-4 | |
Recombinant protein fragment corresponding to amino acids 1-128 of human KRTAP2-4 (NP_149440) produced in E.coli. | |
0.1 mL | |
85294.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI3B4 | |
Flow Cytometry | |
High sulfur keratin-associated protein 2.4, KAP2.4MGC119884, keratin associated protein 2-4, Keratin-associated protein 2.4, keratin-associated protein 2-4, KRTAP2.4, MGC119882, MGC119885, MGC74790 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction