Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP2-4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566240 Supplier Invitrogen™ Supplier No. PA566240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVTCQTTVCRP VTCVPRCTRP ICEPC Highest antigen sequence identity to the following orthologs - mouse 92%, rat 88%.

In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP2-4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRTAP2-4
Gene Accession No. Q9BYR9
Gene Alias high sulfur keratin-associated protein 2.4; KAP2.1B; KAP2.3; KAP2.4; keratin associated protein 2.1B; keratin associated protein 2-4; Keratin-associated protein 2.3; Keratin-associated protein 2.4; Keratin-associated protein 2-3; Keratin-associated protein 2-4; KRTAP2.3; KRTAP2.4; KRTAP2-3; KRTAP2-4
Gene Symbols KRTAP2-4
Host Species Rabbit
Immunogen Recombinant Human KRTAP2-4. Recombinant protein control fragment (Product #RP-105634).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.