Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP27-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554438 Supplier Invitrogen™ Supplier No. PA554438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHSHCHSLRS FHNAPPLSAI THGTNPITFE DRLCLPSSFH SRTCFLDNFQ ETCNETTSCQ MTNCEQDLFT DDSCVQSNCF PGVV Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 51%.

In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP27-1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRTAP27-1
Gene Accession No. Q3LI81
Gene Alias KAP27.1; keratin associated protein 27-1; keratin-associated protein 27-1; KRTAP27-1
Gene Symbols KRTAP27-1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRTAP27-1. Recombinant protein control fragment (Product #RP-91804).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 643812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.