Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRTAP3-3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570512
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%; Sheep: 100%.
This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the high sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21.
Specifications
KRTAP3-3 | |
Polyclonal | |
Unconjugated | |
KRTAP3-3 | |
2310020D06Rik; high sulfur keratin-associated protein 3.3; KAP3.3; keratin associated protein 3-2; keratin associated protein 3-3; keratin-associated protein 3.3; keratin-associated protein 3-2; Keratin-associated protein 3-3; KRTAP3.3; Krtap3-2; krtap3-3; Unknown (protein for MGC:139935) | |
Rabbit | |
Affinity chromatography | |
RUO | |
66380 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9D7P0 | |
KRTAP3-3 | |
synthetic peptide directed towards the middle region of human KRTAP3-3 (aa 10-59). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction