Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP3-3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560190 Supplier Invitrogen™ Supplier No. PA560190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HTVWLLEPTC CDNCPPPCHI PQPCV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the high sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP3-3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRTAP3-3
Gene Accession No. Q9BYR6
Gene Alias 2310020D06Rik; high sulfur keratin-associated protein 3.3; KAP3.3; keratin associated protein 3-2; keratin associated protein 3-3; keratin-associated protein 3.3; keratin-associated protein 3-2; Keratin-associated protein 3-3; KRTAP3.3; Krtap3-2; krtap3-3; Unknown (protein for MGC:139935)
Gene Symbols KRTAP3-3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRTAP3-3. Recombinant protein control fragment (Product #RP-99270).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85293
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.