Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP8-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144153
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144153 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144153 Supplier Invitrogen™ Supplier No. PA5144153
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRTAP8-1 Polyclonal Antibody for ELISA, IHC (P)

KRTAP8-1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP8-1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene KRTAP8-1
Gene Accession No. Q8IUC2
Gene Alias AI326176; Component 0.62; HGT; HGT keratin; HGT keratin (AA 1-62); HGT keratin component F; HGTp type I F; HGTpI F; high glycine-tyrosine keratin-associated protein 8.1; High-glycine/tyrosine protein keratin; KAP8; KAP8.1; keratin associated protein 8; keratin associated protein 8-1; keratin associated protein 8-1-like; Keratin, high-tyrosine matrix protein; keratin-associated protein 8.1; keratin-associated protein 8-1; keratin-associated protein KAP8-1; KRTAP8.1; Krtap8-1; LOC114118004
Gene Symbols KRTAP8-1
Host Species Rabbit
Immunogen Recombinant Human Keratin-associated protein 8-1 protein (1-63AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 337879
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.