Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRTAP8-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569396 Supplier Invitrogen™ Supplier No. PA569396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 92%; Rat: 93%; Sheep: 100%.

KRTAP8-1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRTAP8-1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KRTAP8-1
Gene Accession No. Q8IUC2
Gene Alias AI326176; Component 0.62; HGT; HGT keratin; HGT keratin (AA 1-62); HGT keratin component F; HGTp type I F; HGTpI F; high glycine-tyrosine keratin-associated protein 8.1; High-glycine/tyrosine protein keratin; KAP8; KAP8.1; keratin associated protein 8; keratin associated protein 8-1; keratin associated protein 8-1-like; Keratin, high-tyrosine matrix protein; keratin-associated protein 8.1; keratin-associated protein 8-1; keratin-associated protein KAP8-1; KRTAP8.1; Krtap8-1; LOC114118004
Gene Symbols KRTAP8-1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KRTAP8-1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 337879
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.