Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KSHV K8 Antibody (8C12G10G1), Novus Biologicals™
SDP

Catalog No. NB1002189 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002189 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002189 Supplier Novus Biologicals Supplier No. NB1002189
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KSHV K8 Monoclonal specifically detects KSHV K8 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen KSHV K8
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone 8C12G10G1
Conjugate Unconjugated
Dilution Western Blot 1: 500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Gene Alias human herpes virus 8; Kaposi and #039;s sarcoma herpes virus
Host Species Mouse
Immunogen Ni-NTA purified recombinant human KSHV K8 (isoform 'A) expressed in E. Coli strain BL21 (DE3).
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity KSHV K8 (8C12G10G1)
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.