Learn More
Invitrogen™ KSHV K8a Monoclonal Antibody (8C12G10G1)

Description
MA5-14768 targets KSHV K8a in indirect ELISA, IHC and WB applications and shows reactivity with Virus samples. The MA5-14768 immunogen is purified recombinant fragment of KSHV K8a expressed in E. Coli.
Specifications
Specifications
| Antigen | KSHV K8a |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 8C12G10G1 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene Alias | Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus |
| Host Species | Mouse |
| Immunogen | Purified recombinant fragment of KSHV K8 alpha expressed in E. Coli. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.