Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KSHV LANA2 Monoclonal Antibody (CM-A807)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA116663
Description
This antibody cross-reacts with human and mouse viral KSHV.
LANA2 (latency-associated nuclear antigen) is an inhibitor of p53-induced transcription in reporter assays and may be an apoptosis antagonist. It is a nuclear protein associated with Kaposi's sarcoma-associated herpes virus (KSHV), or human herpes virus 8. It is found in abundance in the nuclei of cultured cells from KSHV-infected hematopoietic tissues, such as PEL (primary effusion lymphoma) and CD (multicentric Castleman's disease). It does not appear to be expressed in KS (Kaposi's sarcoma) lesions.
Specifications
KSHV LANA2 | |
Monoclonal | |
2 mg/mL | |
PBS with 0.1% sodium azide | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
CM-A807 | |
Unconjugated | |
LANA 2; LANA-2 | |
Recombinant LANA2 fusion protein. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction