Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KSHV ORF26 Monoclonal Antibody (2F6B8)
GREENER_CHOICE

Catalog No. PIMA515742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515742 Supplier Invitrogen™ Supplier No. MA515742
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15742 targets KSHV ORF26 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15742 immunogen is purified recombinant fragment of KSHV ORF26 expressed in E. Coli.

Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi's sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman's disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. KSHV ORF26 open reading frames are juxtaposed and span from nucleotide (nt) 46933 to 47850 of the virus genome. Akula et al (2003) report it acts as a Capsid protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KSHV ORF26
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F6B8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene Alias Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus
Host Species Mouse
Immunogen Purified recombinant fragment of KSHV ORF26 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.