Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KSHV ORF45 Monoclonal Antibody (2D4A5)
GREENER_CHOICE

Catalog No. PIMA514769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514769 Supplier Invitrogen™ Supplier No. MA514769
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14769 targets KSHV ORF45 in indirect ELISA, IHC and WB applications and shows reactivity with Virus samples. The MA5-14769 immunogen is purified recombinant fragment of KSHV ORF45 expressed in E. Coli.

Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi's sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman's disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. ORF45 protein encoded by an immediate-early gene in the KSHV genome is characterized as a phosphorylated protein, and it is localized in the cytoplasm of infected cells. Studies have shown that ORF45 Protein interacted with cellular IRF-7 and blocked virus-mediated phosphorylation and nuclear translocation of IRF-7. In consequence, ORF45 efficiently inhibited virus-induced production of type I IFN. Zhu et al (2003) reported that the ORF45 Protein was associated with purified virions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KSHV ORF45
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2D4A5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene Alias Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus
Host Species Mouse
Immunogen Purified recombinant fragment of KSHV ORF45 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.