Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KSHV ORF57 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991960
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1960 100 μg
50-199-1961 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1960 Supplier Rockland Immunochemicals Supplier No. 600401A94
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human herpesvirus 8 (KSHV ORF57) protein. A BLAST analysis was used to suggest cross-reactivity with ORF57 protein from human herpesvirus 8 types P and M sources based on 94% homology with the immunizing sequence. Reactivity against homologues from other sources is unknown.

ORF57 (also known as MTA), one of the earliest Kaposi's sarcoma-associated herpesvirus (KSHV) regulatory proteins to be expressed, is essential for virus lytic replication. A counterpart is present in every herpesvirus sequenced, indicating the importance of this signature viral protein, and those examined act post-transcriptionally, affecting RNA splicing and transport. KSHV ORF57 is capable of establishing both lytic and latent replication cycles. In KS, the virus localizes to tumor progenitor endothelial cells, most of which are latently infected. In cell culture, KSHV replication is generally studied using B-cell lines, such as BCBL-1, generated from primary effusion lymphoma material. Most BCBL-1 cells are latently infected, although there is some spontaneous virus reactivation. Addition of chemical inducers such as sodium n-butyrate, 12-O-tetradecanoylphorbol-13-acetate (TPA), and valproic acid (VA) to these cells efficiently induces the lytic cycle and produces virions. KSHV ORF57 protein predominantly localizes to the nucleus and can shuttle between the nucleus and cytoplasm. Most HSV-1 genes are unspliced; by contrast, ORF57 is spliced gene; the protein is 455 amino acids in length and 50kDa in size.

Specifications

Antigen KSHV ORF57
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.55 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.