Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KSHV ORF62 Antibody (5B7B6), Novus Biologicals™

Mouse Monoclonal Antibody
$499.50
Specifications
Antigen | KSHV ORF62 |
---|---|
Clone | 5B7B6 |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
KSHV ORF62 Monoclonal specifically detects KSHV ORF62 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KSHV ORF62 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
Monoclonal | |
Ascites | |
RUO | |
Ascites with 0.03% Sodium Azide | |
4961461 | |
IgM | |
Unpurified |
5B7B6 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Virus | |
Kaposi's Sarcoma Virus, Kaposi's sarcoma-associated herpesvirus | |
Purified recombinant fragment of human KSHV ORF62 expressed in E. Coli. | |
Primary | |
KSHV |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title