Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KSHV ORF62 Monoclonal Antibody (5B7B6)
GREENER_CHOICE

Catalog No. PIMA514770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514770 Supplier Invitrogen™ Supplier No. MA514770
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14770 targets KSHV ORF62 in indirect ELISA, IHC and WB applications and shows reactivity with Virus samples. The MA5-14770 immunogen is purified recombinant fragment of human KSHV ORF62 expressed in E. Coli.

Kaposi's sarcoma-associated herpesvirus (KSHV) belongs to the gamma-(2)-herpesvirus subfamily and has been closely linked to the Kaposi's sarcoma, primary effusion lymphoma (PEL) and multicentric Castleman's disease. The genome of KSHV is 165-170 kb and contains at least 88 open reading frames. At least five major proteins are likely to be involved in the assembly of the HHV-8 capsid, including a protease (encoded by ORF17), the major capsid protein (encoded by ORF25), and three other smaller capsid proteins (encoded by ORF62, ORF26, and ORF65). Previous structural studies have shown that the HSV-1 triplex is a monomer of VP19c and a dimer of VP23 and that the HCMV triplex is similarly composed of a monomer and a dimer. By analogy, the KSHV triplexes are likely also composed of a monomer of the ORF62 protein and a dimer of the ORF26 protein, which are the respective homologs of VP19c and VP23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KSHV ORF62
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5B7B6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene Alias Kaposi's Sarcoma Virus; Kaposi's sarcoma-associated herpesvirus
Host Species Mouse
Immunogen Purified recombinant fragment of human KSHV ORF62 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.